Learn More
STERICYCLE 1.4 Quart RCRA Container
Catalog No. NC2742092
Encompass
Designed for the safe collection, containment and segregation of hazardous pharmaceutical waste to comply with the Resource Conservation and Recovery Act (RCRA). Distinctive black containers are easy to distinguish from other containers. Case of 20 containers.Dimensions: 3.5"x3.5"x10.25"Opening Type: Screw-on CapBrand: CovidienCount: 20 Containers per CaseCapacity: 1.5 QuartLid: WhiteCase Code: 8601RCCaseCase: Case
Catalog No. NC2742092 Supplier STERICYCLE Supplier No. 8601RCC
Designed for the safe collection, containment and segregation of hazardous pharmaceutical waste to comply with the Resource Conservation and Recovery Act (RCRA). Distinctive black containers are easy to distinguish from other containers. Case of 20 containers.Dimensions: 3.5"x3.5"x10.25"Opening Type: Screw-on CapBrand: CovidienCount: 20 Containers per CaseCapacity: 1.5 QuartLid: WhiteCase Code: 8601RCCaseCase: Case