Learn More
New Jersey Fluid Systems LLC 1/4" QUICK CONNECT BODY W/FEM
Supplier:  New Jersey Fluid Systems LLC SSQC4B4PF
Encompass
316 Stainless Steel Swagelok QC4 in. series Quick Connect Body; 1/4 in. female NPT end; Viton o-ring; 3,000psi @100F
Catalog No. NC0575877