Learn More
Chemglass Life Sciences Chemglass Excelon PVC Laboratory Tubing, 1/4 in ID x 7/16 in OD, 50 ft Roll (Standard wall)
SDP
Catalog No. NC2323632
Encompass_Preferred
Chemglass Excelon PVC laboratory tubing is a clear, flexible, standard-wall tubing designed for reliable liquid and gas transfer in laboratory settings. It features a smooth inner bore to reduce buildup, compatibility with autoclaving and repeated gamma sterilization, and good flexibility and durability for routine use.
  • 1/4 in inner diameter.
  • 7/16 in outer diameter.
  • 3/32 in wall thickness.
  • 50 ft per roll.
  • PVC material with Shore A hardness 55.
  • Temperature range -58°F to 165°F.
  • Meets FDA (CFR 21, Part 175.300).
  • Smooth inner bore reduces buildup and simplifies cleaning.
Catalog No. NC2323632 Supplier Chemglass Life Sciences Supplier No. CG730007
Chemglass Excelon PVC laboratory tubing is a clear, flexible, standard-wall tubing designed for reliable liquid and gas transfer in laboratory settings. It features a smooth inner bore to reduce buildup, compatibility with autoclaving and repeated gamma sterilization, and good flexibility and durability for routine use.
  • 1/4 in inner diameter.
  • 7/16 in outer diameter.
  • 3/32 in wall thickness.
  • 50 ft per roll.
  • PVC material with Shore A hardness 55.
  • Temperature range -58°F to 165°F.
  • Meets FDA (CFR 21, Part 175.300).
  • Smooth inner bore reduces buildup and simplifies cleaning.