Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione 98.0+%, TCI America™
SDP

Catalog No. B338225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B338225G 25 g
B3382100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B338225G Supplier TCI America Supplier No. B338225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70356-09-1
Molecular Formula C20H22O3
Molecular Weight (g/mol) 310.39
MDL Number MFCD00210252
InChI Key XNEFYCZVKIDDMS-UHFFFAOYSA-N
PubChem CID 51040
IUPAC Name 1-(4-tert-butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione
SMILES COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 25 g
Formula Weight 310.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.