Learn More
Chemglass Life Sciences 1/4" to 1/2" PFA Compression Fitting
SDP
Catalog No. CG197520
Encompass_Preferred
1/4" to 1/2" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.
Catalog No. CG197520 Supplier Chemglass Life Sciences Supplier No. CG197520
1/4" to 1/2" PFA Compression Fitting. Designed for use with our CG-1975 heavy wall PTFE tubing. Fittings are made entirely of chemically inert corrosion resistant PFA PTFE. Sizes and configurations other than those listed are available.