Learn More
New Jersey Fluid Systems LLC 1/4" TUBE STB X 1/4"HOSE BARB
Catalog No. NC0575880
Encompass
316 Stainless Steel Swagelok Hose Connector x Tube Adapter. 1/4 in. Hose Barb x 1/4 in. OD Tube Stub adapter.
Catalog No. NC0575880 Supplier New Jersey Fluid Systems LLC Supplier No. SS4HCA401
316 Stainless Steel Swagelok Hose Connector x Tube Adapter. 1/4 in. Hose Barb x 1/4 in. OD Tube Stub adapter.