Learn More
Ace Glass, Inc. J thermocouple sensor, 7mm OD x 12in PTFE covered Inconel sheath, detachable lead sold separately
SDP
Catalog No. 50977508
Encompass
Catalog No. 50-977-508 Supplier Ace Glass, Inc. Supplier No. 1214125