Learn More
Ace Glass, Inc. K thermocouple sensor, 7mm OD x 48in PTFE covered Inconel sheath, detachable lead sold separately
SDP
Catalog No. 50977519
Encompass
Catalog No. 50-977-519 Supplier Ace Glass, Inc. Supplier No. 1214159