Learn More
Accuflow Systems Sani-Tech Peristaltic Tubing, Platinum Silicone, 0.250 ID, 0.375 OD
Catalog No. NC3246161
Encompass_Preferred
Sani-Tech SPT-60L peristaltic pump tubing is a platinum-cured silicone tubing engineered for pharmaceutical final-fill and precision peristaltic pumping. It delivers low spallation, extended pump life, and accurate, consistent dispensing suitable for single-use assemblies and biopharmaceutical manufacturing.
  • Platinum-cured silicone material.
  • Designed for final fill and precision peristaltic pumping.
  • Low spallation and extended pump life.
  • 0.250 in inner diameter, 0.375 in outer diameter, 0.063 in wall thickness.
  • Burst pressure approximately 64 psi (4.4 bar).
  • Full extractables report and validation package available.
Catalog No. NC3246161 Supplier Accuflow Systems Supplier No. SPT60L2502
Sani-Tech SPT-60L peristaltic pump tubing is a platinum-cured silicone tubing engineered for pharmaceutical final-fill and precision peristaltic pumping. It delivers low spallation, extended pump life, and accurate, consistent dispensing suitable for single-use assemblies and biopharmaceutical manufacturing.
  • Platinum-cured silicone material.
  • Designed for final fill and precision peristaltic pumping.
  • Low spallation and extended pump life.
  • 0.250 in inner diameter, 0.375 in outer diameter, 0.063 in wall thickness.
  • Burst pressure approximately 64 psi (4.4 bar).
  • Full extractables report and validation package available.