Learn More
Cayman Chemical 1-(4-Chlorobenzhydryl)piperazine, Purity: ≥98%, CAS: 303-26-4, 10 g
SDP
Catalog No. 502950301
Encompass_Preferred
An inactive metabolite of meclizine and chlorcyclizine; has been found as an impurity in commercial preparations of hydroxyzine and cetirizine; has been used in the synthesis of Nav1.7 inhibitors, anticancer agents, and antihistamines
Catalog No. 50-295-0301 Supplier Cayman Chemical Supplier No. 3451810 G
An inactive metabolite of meclizine and chlorcyclizine; has been found as an impurity in commercial preparations of hydroxyzine and cetirizine; has been used in the synthesis of Nav1.7 inhibitors, anticancer agents, and antihistamines