Learn More
Cayman Chemical 1 4DIbenzylpIperazInhydro 10mg
SDP
Catalog No. 502951220
Encompass_Preferred
A common impurity produced as a reaction byproduct during the synthesis of BZP; intended for research and forensic applications
Catalog No. 50-295-1220 Supplier Cayman Chemical Supplier No. 1120610 MG
A common impurity produced as a reaction byproduct during the synthesis of BZP; intended for research and forensic applications