Learn More
MSC CDI 1501MRMH 1/4" Drive Micrometer Torque Wrench 20 to 150 In/Lb, 0.5 In/Lb Graduation, 10-5/32" Overall Length, Ratcheting Head with Reverse Lever
Catalog No. 501295310
Encompass
CDI 1501MRMH 1/4" Drive Micrometer Torque Wrench 20 to 150 In/Lb, 0.5 In/Lb Graduation, 10-5/32" Overall Length, Ratcheting Head with Reverse Lever
Catalog No. 50-129-5310 Supplier MSC Supplier No. 67557744
CDI 1501MRMH 1/4" Drive Micrometer Torque Wrench 20 to 150 In/Lb, 0.5 In/Lb Graduation, 10-5/32" Overall Length, Ratcheting Head with Reverse Lever