Learn More
AVANTOR MASTERFLEX (VWR LLC) Rubberfab TR-EXT-.25-SET Torque Tee Sanitary Clamp Securing Device Drive Extension Set, 1/4"", 3 pieces
Catalog No. NC1276759
Encompass
Use the Torque Tee to install and remove sanitary clamps safely and securely, even in hard-to-reach spaces. The shape of the handle provides plenty of leverage to reach the factory preset torque. Just listen for the audible click to know the clamp wing nut is properly tightened. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle.
Catalog No. NC1276759 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX2350804
Use the Torque Tee to install and remove sanitary clamps safely and securely, even in hard-to-reach spaces. The shape of the handle provides plenty of leverage to reach the factory preset torque. Just listen for the audible click to know the clamp wing nut is properly tightened. If you need to loosen or remove the clamp, simply disengage the ratcheting mechanism to lock the handle.