Learn More
MSC Parker Hydraulic Coupler, Stainless Steel, 1/4 in NPTF, 2000 psi
Catalog No. NC3595418
Encompass
This hydraulic hose female pipe rigid coupler fitting is constructed from stainless steel with NPTF threading. It is designed for 1/4 in hose and 1/4 in threads, rated for 2,000 psi, and features a straight, threaded connection. It is suitable for diverse applications including gases, industrial hydraulic lines, water and coolant lines, mobile equipment, and food and chemical processing, complying with ISO 7241-1, Series B standards.
  • Constructed from stainless steel
  • Designed for 1/4 in hose and 1/4 in NPTF thread
  • Rated for 2,000 psi with maximum working pressure of 5,000 psi
  • Straight, threaded connection
  • Suitable for gases, industrial hydraulic lines, water and coolant lines, mobile equipment, and food and chemical processing
  • Complies with ISO 7241-1, Series B standards
  • Operates within a temperature range of -15°F to 400°F
Catalog No. NC3595418 Supplier MSC Supplier No. 33838863
This hydraulic hose female pipe rigid coupler fitting is constructed from stainless steel with NPTF threading. It is designed for 1/4 in hose and 1/4 in threads, rated for 2,000 psi, and features a straight, threaded connection. It is suitable for diverse applications including gases, industrial hydraulic lines, water and coolant lines, mobile equipment, and food and chemical processing, complying with ISO 7241-1, Series B standards.
  • Constructed from stainless steel
  • Designed for 1/4 in hose and 1/4 in NPTF thread
  • Rated for 2,000 psi with maximum working pressure of 5,000 psi
  • Straight, threaded connection
  • Suitable for gases, industrial hydraulic lines, water and coolant lines, mobile equipment, and food and chemical processing
  • Complies with ISO 7241-1, Series B standards
  • Operates within a temperature range of -15°F to 400°F