Learn More
Waters Corp Waters Compression Screw, Short, 1/4 in., Gold-plated
Catalog No. NC9377713 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This compression screw is designed for use in various chromatography systems. It features a short, 1/4 in. gold-plated design, ensuring reliable performance.
  • Fitting type
  • Short length
  • 1/4 in. size
  • Gold-plated for enhanced durability
  • Supplied in packs of 10
  • Compatible with UPC2, SFC, HPLC, and UPLC systems
Catalog No. NC9377713 Supplier Waters Corp Supplier No. 700002634
This compression screw is designed for use in various chromatography systems. It features a short, 1/4 in. gold-plated design, ensuring reliable performance.
  • Fitting type
  • Short length
  • 1/4 in. size
  • Gold-plated for enhanced durability
  • Supplied in packs of 10
  • Compatible with UPC2, SFC, HPLC, and UPLC systems