Learn More
Cayman Chemical 1-(4-Methoxyphenyl)piperazine (hydrochloride), Purity: ≥98%, CAS: 38869-47-5, 10 mg
SDP
Catalog No. 502954316
Encompass_Preferred
A piperazine derivative assumed to have a mixed mechanism of action, with serotonergic and dopamine antagonistic properties in a similar fashion to 3,4-methylenedioxymethamphetamine; intended for research and forensic purposes
Catalog No. 50-295-4316 Supplier Cayman Chemical Supplier No. 1402010 MG
A piperazine derivative assumed to have a mixed mechanism of action, with serotonergic and dopamine antagonistic properties in a similar fashion to 3,4-methylenedioxymethamphetamine; intended for research and forensic purposes