Learn More
JDM LAB SOLUTIONS LLC 1.4ml external thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 2019131
Encompass
1 4ml 2D barcoded external thread storage tube with 1D linear barcode human readable tube ID SBS rack format pre-capped with white screw cap Sterile
Catalog No. 50-000-14225