Learn More
JDM LAB SOLUTIONS LLC 1.4ml external thread 2D Tube
Supplier: JDM LAB SOLUTIONS LLC 2019121
1 4ml 2D barcoded external thread storage tube with 1D linear barcode human readable tube ID SBS rack format pre-capped with blue screw cap Sterile