Learn More
JDM LAB SOLUTIONS LLC 1.4ml external thread 2D Tube
Catalog No. 5000014317
Encompass
1 4ml 2D barcoded external thread storage tube SBS rack format precapped with BLUE screw cap sterile
Catalog No. 50-000-14317 Supplier JDM LAB SOLUTIONS LLC Supplier No. 2010911
1 4ml 2D barcoded external thread storage tube SBS rack format precapped with BLUE screw cap sterile