Learn More
JDM LAB SOLUTIONS LLC 1.4ml external thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 2010901
Encompass
1 4ml 2D barcoded external thread storage tube SBS rack format precapped with WHITE screw cap sterile
Catalog No. 50-000-14212