Learn More
JDM LAB SOLUTIONS LLC 1.4ml Internal thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 2010521
Encompass
1 4ml 2D barcoded internal thread storage tube SBS rack format precapped with BLUE screw cap sterile
Catalog No. 50-000-14246