Learn More
MSC 1/4 Npt Brass, Quick Disconnect, Valved Coupling Insert
Supplier:  MSC 88203864
Encompass
250 Max psi, -40 to 180°F, 1.22" OAL x 0.49" Overall Height, Chrome Plated
Catalog No. NC1632782