Learn More
Ace Glass, Inc. PTFE 1/4in tube compression fitting to 1/8in male NPT
SDP
Catalog No. 50977670
Encompass
1/4 PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-670 Supplier Ace Glass, Inc. Supplier No. 1270908
1/4 PTFE tube compression fitting to 1/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.