Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, 0.250 in ID, 0.438 in OD, Platinum-Cured
Catalog No. NC3246164
Encompass_Preferred
Sani-Tech SPT-60L is a platinum-cured silicone tubing engineered for high-accuracy peristaltic pumping and final-fill operations. It offers low spallation, excellent dimensional stability, and long pump life, making it suitable for biopharmaceutical manufacturing and single-use assemblies.
  • 0.250 in (6.4 mm) inner diameter.
  • 0.438 in (11.1 mm) outer diameter.
  • 0.094 in (2.4 mm) wall thickness.
  • Platinum-cured silicone for low spallation and long pump life.
  • Intended for peristaltic pumping, final fill, and single-use assemblies.
Catalog No. NC3246164 Supplier Accuflow Systems Supplier No. SPT60L2503
Sani-Tech SPT-60L is a platinum-cured silicone tubing engineered for high-accuracy peristaltic pumping and final-fill operations. It offers low spallation, excellent dimensional stability, and long pump life, making it suitable for biopharmaceutical manufacturing and single-use assemblies.
  • 0.250 in (6.4 mm) inner diameter.
  • 0.438 in (11.1 mm) outer diameter.
  • 0.094 in (2.4 mm) wall thickness.
  • Platinum-cured silicone for low spallation and long pump life.
  • Intended for peristaltic pumping, final fill, and single-use assemblies.