Learn More
Medchemexpress LLC 1-[5-(2,4-difluorophenyl)sulfanyl-4-nitrothiophen-2-yl]ethanone | 1247819-59-5 | MFCD22580421 | ≥98.0% | 315.32 g·mol-1 | C12H7F2NO3S2 | 5 MG
SDP
Catalog No. 5000222915
Encompass_Preferred
A cell-permeable small-molecule inhibitor of the deubiquitinating enzyme ubiquitin-specific protease 7 (USP7) that also inhibits USP47. It is used in biochemical and cell-based studies to probe deubiquitination pathways; reported EC50 values are 8.01 μM for USP7 and 8.74 μM for USP47. Molecular formula C12H7F2NO3S2; molecular weight 315.32 g·mol-1; CAS 1247819-59-5.
  • Cell-permeable small-molecule inhibitor of USP7 and USP47 (EC50 8.01 μM and 8.74 μM).
  • Suitable for biochemical and cell-based deubiquitination assays.
  • Reported activity values support use in target validation and pathway studies.
  • Provided in small research quantities for laboratory experiments (1-10 mg).
  • Molecular formula C12H7F2NO3S2 and molecular weight 315.32 g·mol-1.
Catalog No. 50-002-22915 Supplier Medchemexpress LLC Supplier No. HY138655MG
A cell-permeable small-molecule inhibitor of the deubiquitinating enzyme ubiquitin-specific protease 7 (USP7) that also inhibits USP47. It is used in biochemical and cell-based studies to probe deubiquitination pathways; reported EC50 values are 8.01 μM for USP7 and 8.74 μM for USP47. Molecular formula C12H7F2NO3S2; molecular weight 315.32 g·mol-1; CAS 1247819-59-5.
  • Cell-permeable small-molecule inhibitor of USP7 and USP47 (EC50 8.01 μM and 8.74 μM).
  • Suitable for biochemical and cell-based deubiquitination assays.
  • Reported activity values support use in target validation and pathway studies.
  • Provided in small research quantities for laboratory experiments (1-10 mg).
  • Molecular formula C12H7F2NO3S2 and molecular weight 315.32 g·mol-1.