Learn More
Medchemexpress LLC 1-{5-[(2,4-Dimethylphenoxy)methyl]-1,3,4-thiadiazol-2-yl}-3-(4-methylphenyl)urea | 428478-94-8 | 98.1% | 368.45 g/mol | C19H20N4O2S | 5 MG
SDP
Catalog No. 5000262308
Encompass_Preferred
cyt-PTPε Inhibitor-1 is a small-molecule research reagent that potently inhibits cytosolic protein tyrosine phosphatase epsilon (cyt-PTPε). It binds the enzyme's catalytic domain, prevents c-Src dephosphorylation and activation, and has reported anti-osteoclastic activity. Supplied as a white to off-white solid with high purity for laboratory use.
  • Potent cytosolic protein tyrosine phosphatase epsilon inhibitor.
  • Binds the catalytic domain and blocks c-Src activation.
  • Reported anti-osteoclastic biological activity.
  • White to off-white solid suitable for laboratory handling.
  • Molecular formula C19H20N4O2S; molecular weight 368.45 g/mol.
  • Intended for research use only.
Catalog No. 50-002-62308 Supplier Medchemexpress LLC Supplier No. HY1128005MG
cyt-PTPε Inhibitor-1 is a small-molecule research reagent that potently inhibits cytosolic protein tyrosine phosphatase epsilon (cyt-PTPε). It binds the enzyme's catalytic domain, prevents c-Src dephosphorylation and activation, and has reported anti-osteoclastic activity. Supplied as a white to off-white solid with high purity for laboratory use.
  • Potent cytosolic protein tyrosine phosphatase epsilon inhibitor.
  • Binds the catalytic domain and blocks c-Src activation.
  • Reported anti-osteoclastic biological activity.
  • White to off-white solid suitable for laboratory handling.
  • Molecular formula C19H20N4O2S; molecular weight 368.45 g/mol.
  • Intended for research use only.