Learn More
Azzota Corp Hollow Cathode Lamp (Hcl) Nickel, 1.5 In. (37mm) Fits: Varian, Analytic Jena, Gbc, Hitachi , Shimadzu , Tja, Thermo Unicam, Thermo Elemental, Buck, Phillips, Leeman Labs, Chemito
Catalog No. 507532929
Encompass_Preferred
Hollow Cathode Lamp (HCL) Nickel, 1.5 in. (37mm) diameter fit in a wide range of instruments:- Varian, Analytic Jena, GBC, Hitachi , Shimadzu , TJA, Thermo Unicam, Thermo Elemental, Buck, Phillips, Leeman Labs, Chemito and many others
Catalog No. 50-753-2929 Supplier Azzota Corp Supplier No. SHC15HCL
Hollow Cathode Lamp (HCL) Nickel, 1.5 in. (37mm) diameter fit in a wide range of instruments:- Varian, Analytic Jena, GBC, Hitachi , Shimadzu , TJA, Thermo Unicam, Thermo Elemental, Buck, Phillips, Leeman Labs, Chemito and many others