Learn More
Medchemexpress LLC 1-[5-[(4,5-diphenyl-1H-imidazol-2-yl)thio]pentyl]-3,5-dimethyl-1H-pyrazole | 136609-26-2 | 99.7% | 416.6 g·mol⁻¹ | C25H28N4S | 100 MG
SDP
Catalog No. 5000248976
Encompass_Preferred
RP 70676 is a small-molecule research compound that potently inhibits acyl-CoA:cholesterol O-acyltransferase (ACAT). It has demonstrated low-nanomolar activity in rodent enzyme assays and has been used in preclinical pharmacology studies.
  • Potent ACAT inhibition with reported IC50 of 25 nM (rat) and 44 nM (rabbit).
  • High purity suitable for research use (vendor-reported values ≥98-99.7%).
  • Reported oral bioavailability in animal studies.
  • Molecular formula C25H28N4S and molecular weight 416.6 g·mol⁻¹.
  • Supplied with commercial characterization data and literature references for preclinical research.
Catalog No. 50-002-48976 Supplier Medchemexpress LLC Supplier No. HY101576100MG
RP 70676 is a small-molecule research compound that potently inhibits acyl-CoA:cholesterol O-acyltransferase (ACAT). It has demonstrated low-nanomolar activity in rodent enzyme assays and has been used in preclinical pharmacology studies.
  • Potent ACAT inhibition with reported IC50 of 25 nM (rat) and 44 nM (rabbit).
  • High purity suitable for research use (vendor-reported values ≥98-99.7%).
  • Reported oral bioavailability in animal studies.
  • Molecular formula C25H28N4S and molecular weight 416.6 g·mol⁻¹.
  • Supplied with commercial characterization data and literature references for preclinical research.