Learn More
Medchemexpress LLC 1-(5-(4,5-diphenyl-1H-imidazol-2-ylthio)pentyl)-3,5-dimethyl-1H-pyrazole | 136609-26-2 | 99.4% | 416.58 g·mol⁻1 | C25H28N4S | 50 MG
SDP
Catalog No. 5000248496
Encompass_Preferred
RP 70676 is a small-molecule inhibitor of acyl-CoA:cholesterol O-acyltransferase (ACAT) reported to have low-nanomolar activity in rodent enzymes. It is supplied as a high-purity solid for research use, with molecular formula C25H28N4S and molecular weight 416.58 g·mol⁻1. Reported IC50 values are 25 nM (rat ACAT) and 44 nM (rabbit ACAT).
  • Potent ACAT inhibition with low-nanomolar IC50 values.
  • High purity suitable for biological assays.
  • Solid form, soluble in DMSO for convenient preparation.
  • Supplied with certificate of analysis for batch-specific purity.
  • Suitable for in vitro and in vivo preclinical studies.
Catalog No. 50-002-48496 Supplier Medchemexpress LLC Supplier No. HY10157650MG
RP 70676 is a small-molecule inhibitor of acyl-CoA:cholesterol O-acyltransferase (ACAT) reported to have low-nanomolar activity in rodent enzymes. It is supplied as a high-purity solid for research use, with molecular formula C25H28N4S and molecular weight 416.58 g·mol⁻1. Reported IC50 values are 25 nM (rat ACAT) and 44 nM (rabbit ACAT).
  • Potent ACAT inhibition with low-nanomolar IC50 values.
  • High purity suitable for biological assays.
  • Solid form, soluble in DMSO for convenient preparation.
  • Supplied with certificate of analysis for batch-specific purity.
  • Suitable for in vitro and in vivo preclinical studies.