Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one, 97%, Thermo Scientific™

Catalog No. TL00212DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL00212DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00212DA Supplier Thermo Scientific Chemicals Supplier No. TL00212DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17610-21-8
Molecular Formula C16H22O
Molecular Weight (g/mol) 230.351
InChI Key IHUSZOMIBSDQTB-UHFFFAOYSA-N
Synonym 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethanone, 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethan-1-one, 1,1,4,4-tetramethyl-6-acetyltetralin, 1-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethanone, 1-5,5,8,8-tetramethyl-5,6,7,8-tetrahydro-naphthalen-2-yl-ethanone, 6-acetyl-1,2,3,4-tetrahydro-1,1,4,4-tetramethylnaphthalene, 2-acetyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalene, 6-acetyl-1,1,4,4-tetramethyltetralin, 6-acetyl-1,1,4,4-tetra-methyl-tetralin, 1,1,4,4-tetramethyl-1,2,3,4-tetrahydro-6-acetylnaphthalene
PubChem CID 2747562
IUPAC Name 1-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)C(CCC2(C)C)(C)C

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 230.35
Percent Purity 95%
Chemical Name or Material 1-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.