Learn More
Medchemexpress LLC 1-(5,6-dimethylthieno[2,3-d]pyrimidin-4-yl)piperidine-4-carboxamide | 785705-53-5 | 99.0% | C14H18N4OS | 10MG
SDP
Catalog No. 5000259911
Encompass_Preferred
M4 mAChR agonist-1 (compound 10a) is a research-stage muscarinic M4 receptor agonist reported to have an EC50 >10 μM for human M4. Supplied as a light yellow to brown solid, the compound has molecular formula C14H18N4OS and molecular weight 290.38 g/mol. It is intended for laboratory research use only and should be handled and stored according to the provided safety data sheet and storage recommendations.
Catalog No. 50-002-59911 Supplier Medchemexpress LLC Supplier No. HY14702810MG
M4 mAChR agonist-1 (compound 10a) is a research-stage muscarinic M4 receptor agonist reported to have an EC50 >10 μM for human M4. Supplied as a light yellow to brown solid, the compound has molecular formula C14H18N4OS and molecular weight 290.38 g/mol. It is intended for laboratory research use only and should be handled and stored according to the provided safety data sheet and storage recommendations.