Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5,9-Triazacyclododecane 95.0+%, TCI America™
SDP

Catalog No. T1876100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1876100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1876100MG Supplier TCI America Supplier No. T1876100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 294-80-4
Molecular Formula C9H21N3
Molecular Weight (g/mol) 171.288
InChI Key VQFZKDXSJZVGDA-UHFFFAOYSA-N
PubChem CID 470778
IUPAC Name 1,5,9-triazacyclododecane
SMILES C1CNCCCNCCCNC1

Specifications

Melting Point 36°C
Boiling Point 78°C
Quantity 100 mg
UN Number 3259
Formula Weight 171.29
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5,9-Triazacyclododecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.