Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Anhydro-D-sorbitol, 97%

Catalog No. AC459910010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459910010 1 g
AC459912500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459910010 Supplier Thermo Scientific Chemicals Supplier No. 459910010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 154-58-5 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154-58-5
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
InChI Key MPCAJMNYNOGXPB-SLPGGIOYSA-N
Synonym 1,5-anhydro-d-glucitol, 1,5-anhydroglucitol, 1,5-anhydrosorbitol, 1,5-anhydro-d-sorbitol, 1-deoxy-d-glucopyranose, aceritol, 1-deoxy-d-glucose, d-glucitol, 1,5-anhydro, unii-54bb3b7xmz, glucitol, 1,5-anhydro
PubChem CID 64960
ChEBI CHEBI:16070
IUPAC Name (2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 141.0°C to 144.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 1,5-Anhydro-D-sorbitol
EINECSNumber 205-829-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.