Learn More
Azzota Corp 1.5"" Gold - Au Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 501612958
Encompass_Preferred
1.5" Gold - Au Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 50-161-2958 Supplier Azzota Corp Supplier No. SHC15AU
1.5" Gold - Au Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm