Learn More
Medchemexpress LLC 1-((5-chloro-1H-indol-2-yl)methyl)-N,N-dimethylmethanamine | 1210-81-7 | 99.7% | 236.74 g·mol⁻¹ | C13H17ClN2 | 50 MG
SDP
Catalog No. 5000246329
Encompass_Preferred
ST1936 is a selective 5-HT6 receptor agonist provided for laboratory research use. It is reported at 99.70% purity, has molecular formula C13H17ClN2, molecular weight 236.74 g·mol⁻¹, and CAS 1210-81-7. The compound is supplied in small milligram pack sizes suitable for pharmacology and signaling studies.
  • High purity (99.7%).
  • Selective 5-HT6 receptor agonist.
  • Suitable for in vitro and in vivo studies.
  • Available in small milligram pack sizes.
  • Includes safety data and analytical documentation.
Catalog No. 50-002-46329 Supplier Medchemexpress LLC Supplier No. HY10311050MG
ST1936 is a selective 5-HT6 receptor agonist provided for laboratory research use. It is reported at 99.70% purity, has molecular formula C13H17ClN2, molecular weight 236.74 g·mol⁻¹, and CAS 1210-81-7. The compound is supplied in small milligram pack sizes suitable for pharmacology and signaling studies.
  • High purity (99.7%).
  • Selective 5-HT6 receptor agonist.
  • Suitable for in vitro and in vivo studies.
  • Available in small milligram pack sizes.
  • Includes safety data and analytical documentation.