Learn More
Medchemexpress LLC 1-(5-chloro-2-hydroxyphenyl)-3-[2-chloro-5-(trifluoromethyl)phenyl]urea | 501684-93-1 | 8ge98.0% | 25 MG
SDP
Catalog No. 5000230392
Encompass_Preferred
NS 1738 is a research chemical that acts as a positive allosteric modulator of the α7 nicotinic acetylcholine receptor (α7 nAChR). It potentiates α7 nAChR activity (EC50 = 3.4 μM in oocyte assays) and is provided for laboratory research use only; not for human or clinical use.
  • Positive allosteric modulator of α7 nAChR (EC50 = 3.4 μM in oocyte assays).
  • Reported molecular weight 365.13 g·mol⁻1 and formula C14H9Cl2F3N2O2.
  • CAS registry number 501684-93-1 for substance identification.
  • Typically supplied in small research quantities (e.g., 25 MG) for in vitro studies.
  • Intended for laboratory research use only; not for human use.
Catalog No. 50-002-30392 Supplier Medchemexpress LLC Supplier No. HY1215125MG
NS 1738 is a research chemical that acts as a positive allosteric modulator of the α7 nicotinic acetylcholine receptor (α7 nAChR). It potentiates α7 nAChR activity (EC50 = 3.4 μM in oocyte assays) and is provided for laboratory research use only; not for human or clinical use.
  • Positive allosteric modulator of α7 nAChR (EC50 = 3.4 μM in oocyte assays).
  • Reported molecular weight 365.13 g·mol⁻1 and formula C14H9Cl2F3N2O2.
  • CAS registry number 501684-93-1 for substance identification.
  • Typically supplied in small research quantities (e.g., 25 MG) for in vitro studies.
  • Intended for laboratory research use only; not for human use.