Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(5-Chloro-2-phenoxyphenyl)ethanone

Catalog No. JRD08711G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
JRD08711G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JRD08711G Supplier Sigma Aldrich Supplier No. JRD08711G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-ethynyl-α,α,α-trifluorotoluene reacts with ethanol to afford the corresponding α,α-alkynyl ester
  • 4-ethynyl-α,α,α-trifluorotoluene plays the role of a functionalized alkyne in copper-catalyzed azide-alkyne cycloaddition (cuaac) reaction on brpp thin films.used to synthesize self-assembled monolayers (sams)

Specifications

CAS 70958-18-8
Molecular Formula C14H11ClO2
Linear Formula C14H11ClO2
MDL Number MFCD17677465
Quantity 5 g
Molecular Weight (g/mol) 246.69
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.