Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(5-chloronaphthalen-1-yl)sulfonyl-1,4-diazepane hydrochloride | 105637-50-1 | MFCD00065525 | 99.9% | 361.29 g/mol | C15H18Cl2N2O2S | 100 MG
SDP

Supplier:  Medchemexpress LLC HY100932100MG

Encompass_Preferred

ML-9 is a small-molecule research inhibitor of Akt kinase that also inhibits myosin light-chain kinase (MLCK) and STIM1. It shows reported Ki values of 4 μM (MLCK), 32 μM (PKA), and 54 μM (PKC), and can induce autophagy by stimulating autophagosome formation. Intended for laboratory research use; handle and store under dry, sealed conditions.

  • Inhibits Akt kinase, MLCK, and STIM1.
  • Induces autophagy by stimulating autophagosome formation.
  • High purity: 99.86%.
  • Molecular weight: 361.29 g/mol.
  • Chemical formula: C15H18Cl2N2O2S.
  • Available as solids (including 100 mg) and 10 mM solutions in DMSO.
  • Storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-12647


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.