Learn More
Azzota Corp 1.5" Chromium-Cr Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 501612961
Encompass_Preferred
1.5" Chromium-Cr Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 50-161-2961 Supplier Azzota Corp Supplier No. SHC15CR
1.5" Chromium-Cr Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm