Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,5-Cyclooctadiene

Catalog No. C109207100M Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C109207100M 100 mL
C1092071L 1 L
C1092074L 4 L
C109207500M 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. C109207100M Supplier Sigma Aldrich Supplier No. C109207100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,5-Cyclooctadiene

Specifications

CAS 111-78-4
Melting Point -69°C (lit.)
Density 0.882 g/mL (at 25°C (literature))
Boiling Point 149°C to 150°C (lit.)
Molecular Formula C8H12
Refractive Index n20/D 1.493 (literature)
Linear Formula C8H12
MDL Number MFCD00001752
Quantity 100 mL
Molecular Weight (g/mol) 108.18
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.