Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Cyclooctadiene, 99%, stabilized

Catalog No. p-3736071
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297122500 250 mL Glass bottle
AC297120010 1 L Glass bottle
AC297120025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC297122500 Supplier Thermo Scientific Chemicals Supplier No. 297122500
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 111-78-4 | C8H12 | 108.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-78-4
Molecular Formula C8H12
Molecular Weight (g/mol) 108.18
MDL Number MFCD00001752
InChI Key VYXHVRARDIDEHS-QGTKBVGQSA-N
Synonym 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
PubChem CID 10937607
SMILES C1C\C=C/CC\C=C/1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -69.5°C
Color Colorless
Density 0.8800g/mL
Boiling Point 151°C (101.3 kPa)
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4920 to 1.4940
Quantity 250 mL
Beilstein 05, 116
Specific Gravity 0.88
Merck Index 15, 2730
Solubility Information Solubility in water: 780mg/l (20°C). Other solubilities: soluble in benzene and carbon tetrachloride
IUPAC Name (5Z)-cycloocta-1,5-diene
Stabilizer 50 to 150ppm t-butylcatechol
Viscosity 1.3 mPa.s (20°C)
Formula Weight 108.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 5-Cyclooctadiene, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a
EINECSNumber 203-907-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.