Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (1,5-Cyclooctadiene)Bis(Methyldiphenylphosphine)Iridium(I) Hexafluorophosphate

Catalog No. 33745550MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-745-550MG 50 mg
33-745-5500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-745-550MG Supplier Sigma Aldrich Supplier No. 33745550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(1,5-Cyclooctadiene)Bis(Methyldiphenylphosphine)Iridium(I) Hexafluorophosphate

Specifications

CAS 38465-86-0
Melting Point 224°C
Molecular Formula C34H38F6IrP3
Linear Formula C34H38F6IrP3
MDL Number MFCD00064800
Quantity 50 mg
Synonym [Ir(PMePh2)2COD]+ PF6-; [Ir(cod)(PPh2Me)2]PF6; {Ir(COD)[PCH3(C6H5)2]2}PF6
Molecular Weight (g/mol) 845.79 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.