Learn More
Medchemexpress LLC (1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate | 38465-86-0 | >98.0% | 845.79 | C34H38IrP2.PF6 | 100 MG
SDP
Catalog No. 5000299399
Encompass_Preferred
(1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an organometallic iridium(I) complex containing 1,5-cyclooctadiene and two methyldiphenylphosphine ligands, with hexafluorophosphate as the counterion. It is provided as a high-purity catalyst precursor for homogeneous hydrogenation and related organometallic transformations in research settings.
  • High-purity material suitable for catalytic applications.
  • Serves as a catalyst precursor for hydrogenation and transfer hydrogenation.
  • Soluble in common organic solvents for homogeneous reaction conditions.
  • Hexafluorophosphate counterion contributes to complex stability.
  • Packaged in small quantities appropriate for laboratory use.
Catalog No. 50-002-99399 Supplier Medchemexpress LLC Supplier No. HYW035155100MG
(1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an organometallic iridium(I) complex containing 1,5-cyclooctadiene and two methyldiphenylphosphine ligands, with hexafluorophosphate as the counterion. It is provided as a high-purity catalyst precursor for homogeneous hydrogenation and related organometallic transformations in research settings.
  • High-purity material suitable for catalytic applications.
  • Serves as a catalyst precursor for hydrogenation and transfer hydrogenation.
  • Soluble in common organic solvents for homogeneous reaction conditions.
  • Hexafluorophosphate counterion contributes to complex stability.
  • Packaged in small quantities appropriate for laboratory use.