Learn More
Medchemexpress LLC 1,5-cyclooctadiene bis(methyldiphenylphosphine) iridium(I) hexafluorophosphate | 38465-86-0 | MFCD00064800 | 97.0% | 845.79 | C34H38IrP2·PF6 | 25 MG
SDP
Catalog No. 5000303194
Encompass_Preferred
This organoiridium complex, (1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate (CAS 38465-86-0), is a homogeneous catalyst used in hydrogenation and isomerization reactions. It is supplied as an orange to red solid with a formula weight of 845.79 g/mol and is provided in small research quantities.
  • High catalytic activity in hydrogenation and isomerization reactions.
  • Solid, orange to red appearance facilitates handling and storage.
  • Well-defined molecular weight and composition for reproducible results.
  • Suitable for research and small-scale synthetic applications.
  • Typical purity approximately 97.0%.
Catalog No. 50-003-03194 Supplier Medchemexpress LLC Supplier No. 25MGHYW035155
This organoiridium complex, (1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate (CAS 38465-86-0), is a homogeneous catalyst used in hydrogenation and isomerization reactions. It is supplied as an orange to red solid with a formula weight of 845.79 g/mol and is provided in small research quantities.
  • High catalytic activity in hydrogenation and isomerization reactions.
  • Solid, orange to red appearance facilitates handling and storage.
  • Well-defined molecular weight and composition for reproducible results.
  • Suitable for research and small-scale synthetic applications.
  • Typical purity approximately 97.0%.