Learn More
Medchemexpress LLC (1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate | 38465-86-0 | MFCD00064800 | 97.0% | 845.79 g/mol | C34H38F6IrP3 | 50 MG
SDP
Catalog No. 5000299849
Encompass_Preferred
(1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an organometallic iridium complex used as a homogeneous catalyst in organic synthesis. It is typically supplied as an orange to red solid of high purity and is handled under inert conditions for laboratory-scale catalytic transformations.
  • Used as a homogeneous catalyst for hydrogenation and C-H activation.
  • High purity (about 97.0%) for consistent catalytic performance.
  • Orange to red solid stable when stored under inert gas at low temperature.
  • Molecular weight 845.79 g/mol and formula C34H38F6IrP3 for formulation reference.
  • Supplied in small research-scale quantities suitable for laboratory synthesis.
Catalog No. 50-002-99849 Supplier Medchemexpress LLC Supplier No. HYW03515550MG
(1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an organometallic iridium complex used as a homogeneous catalyst in organic synthesis. It is typically supplied as an orange to red solid of high purity and is handled under inert conditions for laboratory-scale catalytic transformations.
  • Used as a homogeneous catalyst for hydrogenation and C-H activation.
  • High purity (about 97.0%) for consistent catalytic performance.
  • Orange to red solid stable when stored under inert gas at low temperature.
  • Molecular weight 845.79 g/mol and formula C34H38F6IrP3 for formulation reference.
  • Supplied in small research-scale quantities suitable for laboratory synthesis.