Learn More
Medchemexpress LLC (1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate | 38465-86-0 | MFCD00064800 | 97.0% | 845.79 g·mol⁻¹ | C34H38IrP2·PF6 | 250 MG
SDP
Catalog No. 5000303534
Encompass_Preferred
(1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an organometallic iridium(I) complex used as a homogeneous hydrogenation catalyst in organic synthesis. It has the formula C34H38IrP2·PF6, molecular weight 845.79 g·mol⁻¹, and is supplied with a typical purity of 97%.
  • High catalytic activity for hydrogenation reactions.
  • Suitable for homogeneous catalytic processes in organic synthesis.
  • Available in small quantities for research-scale reactions.
  • Stable as a hexafluorophosphate salt for isolation and handling.
  • Supplied with documentation (COA and SDS) for quality and safety.
Catalog No. 50-003-03534 Supplier Medchemexpress LLC Supplier No. 250MGHYW035155
(1,5-Cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an organometallic iridium(I) complex used as a homogeneous hydrogenation catalyst in organic synthesis. It has the formula C34H38IrP2·PF6, molecular weight 845.79 g·mol⁻¹, and is supplied with a typical purity of 97%.
  • High catalytic activity for hydrogenation reactions.
  • Suitable for homogeneous catalytic processes in organic synthesis.
  • Available in small quantities for research-scale reactions.
  • Stable as a hexafluorophosphate salt for isolation and handling.
  • Supplied with documentation (COA and SDS) for quality and safety.