Learn More
Medchemexpress LLC (1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate | 38465-86-0 | MFCD00064800 | 97.0% | 845.79 g/mol | C34H38IrP2·PF6 | 500 MG
SDP
Catalog No. 5000299309
Encompass_Preferred
(1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an organometallic iridium complex used as a homogeneous catalyst and research reagent in organometallic synthesis and hydrogenation studies. Supplied as an orange-red solid with analytical-grade purity, it is intended for laboratory-scale catalytic applications and method development.
  • High purity (97.0%) suitable for research and catalytic studies.
  • Solid, orange to red appearance for straightforward handling and storage.
  • Ir(I) coordination promotes efficient homogeneous catalysis and hydrogenation.
  • PF6 counterion aids solubility in common organic solvents.
  • Available in multiple small-scale pack sizes for laboratory use.
Catalog No. 50-002-99309 Supplier Medchemexpress LLC Supplier No. HYW035155500MG
(1,5-cyclooctadiene)bis(methyldiphenylphosphine)iridium(I) hexafluorophosphate is an organometallic iridium complex used as a homogeneous catalyst and research reagent in organometallic synthesis and hydrogenation studies. Supplied as an orange-red solid with analytical-grade purity, it is intended for laboratory-scale catalytic applications and method development.
  • High purity (97.0%) suitable for research and catalytic studies.
  • Solid, orange to red appearance for straightforward handling and storage.
  • Ir(I) coordination promotes efficient homogeneous catalysis and hydrogenation.
  • PF6 counterion aids solubility in common organic solvents.
  • Available in multiple small-scale pack sizes for laboratory use.