Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,5-Cyclooctadiene{[Dibenzyl((4R,5R)-5-Methyl-2-Phenyl-4,5-Dihydro-4-Oxazolyl)Methyl]Dicyclohexylphosphinite Kappan:Kappap}Iridium(I) Tetrakis(3,5-Bis(Trifluoromethyl)Phenyl)Borate

Catalog No. 71357050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-357-050G 50 g
71-357-0100G 100 g
71-357-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71-357-050G Supplier Sigma Aldrich Supplier No. 71357050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Catalyst for enantioselective hydrogenation of alkenessold in collaboration with Solvias AG

Specifications

CAS 880262-14-6
Molecular Formula C77H70BF24IrNO2P
Linear Formula C77H70BF24IrNO2P
MDL Number MFCD18252665
Quantity 50 g
Synonym Ubaphox; [((4R,5R)-Cy2-Ubaphox)Ir(COD)]BARF
Molecular Weight (g/mol) 1731.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.