Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diamino-4,8-dihydroxyanthraquinone 95.0+%, TCI America™
SDP

Catalog No. D27265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27265G Supplier TCI America Supplier No. D27265G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 145-49-3
Molecular Formula C14H10N2O4
Molecular Weight (g/mol) 270.244
MDL Number MFCD00013722
InChI Key HSYLKWSCFRLSKB-UHFFFAOYSA-N
Synonym 1,5-diamino-4,8-dihydroxyanthraquinone, diaminoanthrarufin, 1,5-diaminoanthrarufin, 4,8-diaminoanthrarufin, 1,5-dihydroxy-4,8-diaminoanthraquinone, anthrarufin, 4,8-diamino, unii-q7yvg56q18, ccris 5599, 4,8-diamino-1,5-dihydroxyanthraquinone, 1,5-diamino-4,8-dihydroxy-anthracene-9,10-dione
PubChem CID 67351
IUPAC Name 1,5-diamino-4,8-dihydroxyanthracene-9,10-dione
SMILES C1=CC(=C2C(=C1N)C(=O)C3=C(C=CC(=C3C2=O)N)O)O

Specifications

Color Black-Purple
Quantity 5 g
Formula Weight 270.24
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diamino-4,8-dihydroxyanthraquinone
EINECSNumber (4)-1269
RTECSNumber CB6480000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.