Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diaminopentane, 98%

Catalog No. AC112320050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112320050 5 mL Glass bottle
AC112320250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC112320050 Supplier Thermo Scientific Chemicals Supplier No. 112320050
Add to Cart
This item is not returnable. View return policy

CAS: 462-94-2 | C5H14N2 | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-94-2
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008239
InChI Key VHRGRCVQAFMJIZ-UHFFFAOYSA-N
Synonym 1,5-diaminopentane, cadaverine, 1,5-pentanediamine, pentamethylenediamine, cadaverin, animal coniine, 1,5-pentamethylenediamine, unii-l90ben6oll, l90ben6oll, n2p
PubChem CID 273
ChEBI CHEBI:18127
IUPAC Name pentane-1,5-diamine
SMILES NCCCCCN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 9°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 178°C to 180°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4570 to 1.463
Linear Formula H2N(CH2)5NH2
Quantity 5 mL
Beilstein 04, 266
Specific Gravity 0.87
Merck Index 15, 1614
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 102.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 5-Diaminopentane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 207-329-0
RTECSNumber SA0200000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.